COM448653819 Combi-Blocks, Inc.

3-Methylimidazo[1,2-a]pyridine-6-carbonitrile

  • MDL number MFCD14559246
  • Cas Number : 952511-46-5
  • Molecular Weight : 157.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0192

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0192
Availability details for
Quantity
  • Availability
  • Add to Cart