COM448653822 Combi-Blocks, Inc.

5-Amino-1-methyl-3-(trifluoromethyl)-1h-pyrazole-4-carboxylic acid

  • MDL number MFCD12827717
  • Cas Number : 317806-51-2
  • Molecular Weight : 209.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0195

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0195
Availability details for
Quantity
  • Availability
  • Add to Cart