COM448653836 Combi-Blocks, Inc.

3-(4-Methoxy-benzyloxy)-phenylamine

  • MDL number MFCD09803253
  • Cas Number : 131770-85-9
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0210

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0210
Availability details for
Quantity
  • Availability
  • Add to Cart