COM448653845 Combi-Blocks, Inc.

4-Hydroxy-2,3,5,6-tetrafluorostyrene

  • MDL number MFCD18074363
  • Cas Number : 385422-30-0
  • Molecular Weight : 192.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0220

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0220
Availability details for
Quantity
  • Availability
  • Add to Cart