COM448653856 Combi-Blocks, Inc.

Methyl 4-(3-hydroxy-3-methylbut-1-ynyl)benzoate

  • MDL number MFCD00168836
  • Cas Number : 33577-98-9
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0231

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0231
Availability details for
Quantity
  • Availability
  • Add to Cart