COM448653862 Combi-Blocks, Inc.

3-Fluoro-4-hydroxy-2-methoxypyridine

  • MDL number MFCD16611067
  • Cas Number : 1227511-87-6
  • Molecular Weight : 143.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0238

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0238
Availability details for
Quantity
  • Availability
  • Add to Cart