COM448653890 Combi-Blocks, Inc.

2-Nitro-5,6,8,9-tetrahydro-benzocyclohepten-7-one

  • MDL number MFCD11869793
  • Cas Number : 740842-50-6
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0267

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0267
Availability details for
Quantity
  • Availability
  • Add to Cart