COM448653891 Combi-Blocks, Inc.

3-[(6-Methylpyrazin-2-yl)oxy]benzoic acid

  • MDL number MFCD09817520
  • Cas Number : 906352-96-3
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0268

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0268
Availability details for
Quantity
  • Availability
  • Add to Cart