COM448653910 Combi-Blocks, Inc.

2-Bromo-3-methyl-4,4,4-trifluorobutyric acid

  • MDL number MFCD08458181
  • Cas Number : 914635-99-7
  • Molecular Weight : 235.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0288

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0288
Availability details for
Quantity
  • Availability
  • Add to Cart