COM448653957 Combi-Blocks, Inc.

6-Maleimido-1-hexanol

  • MDL number MFCD16251432
  • Cas Number : 157503-18-9
  • Molecular Weight : 197.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0338

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0338
Availability details for
Quantity
  • Availability
  • Add to Cart