COM448653966 Combi-Blocks, Inc.

(2-(Trifluoromethyl)pyridin-3-yl)methanol

  • MDL number MFCD11100743
  • Cas Number : 131747-57-4
  • Molecular Weight : 177.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0347

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0347
Availability details for
Quantity
  • Availability
  • Add to Cart