COM448653970 Combi-Blocks, Inc.

L-Beta-homophenylalanine allyl ester hydrochloride

  • MDL number MFCD01863076
  • Cas Number : 352523-25-2
  • Molecular Weight : 255.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0351

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0351
Availability details for
Quantity
  • Availability
  • Add to Cart