COM448653973 Combi-Blocks, Inc.

5-Chloro-1,1,1-trifluoro-3-oxapentane

  • MDL number MFCD00462515
  • Cas Number : 1537-70-8
  • Molecular Weight : 162.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0354

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0354
Availability details for
Quantity
  • Availability
  • Add to Cart