COM448653976 Combi-Blocks, Inc.

1,4-Dioxaspiro[4.5]decan-8-yl methanesulfonate

  • MDL number MFCD09953114
  • Cas Number : 141120-33-4
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0357

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0357
Availability details for
Quantity
  • Availability
  • Add to Cart