COM448653985 Combi-Blocks, Inc.

N-Methyl-n-(3-pyridin-3-ylbenzyl)amine

  • MDL number MFCD07368550
  • Cas Number : 852180-72-4
  • Molecular Weight : 198.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0366

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0366
Availability details for
Quantity
  • Availability
  • Add to Cart