COM448653993 Combi-Blocks, Inc.

Nonyltrimethylammonium bromide

  • MDL number MFCD00041986
  • Cas Number : 1943-11-9
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0374

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0374
Availability details for
Quantity
  • Availability
  • Add to Cart