COM448654008 Combi-Blocks, Inc.

2-(2,4,5-Trifluorophenyl)-4,5-dihydro-4,4-dimethyloxazole

  • MDL number MFCD09037938
  • Cas Number : 125290-72-4
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0389

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0389
Availability details for
Quantity
  • Availability
  • Add to Cart