COM448654026 Combi-Blocks, Inc.

2-Chloro-2-(2'-nitro-4'-chlorophenylhydrazono)acetic acid ethyl ester

  • MDL number MFCD09998338
  • Cas Number : 112091-27-7
  • Molecular Weight : 306.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0408

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0408
Availability details for
Quantity
  • Availability
  • Add to Cart