COM448654035 Combi-Blocks, Inc.

1,2,2-Trichloropentafluoropropane

  • MDL number MFCD00077596
  • Cas Number : 1599-41-3
  • Molecular Weight : 237.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0417

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0417
Availability details for
Quantity
  • Availability
  • Add to Cart