COM448654039 Combi-Blocks, Inc.

Perfluoro(4-methyl-3,6-dioxaoctane)sulfonyl fluoride

  • MDL number MFCD00155782
  • Cas Number : 27744-59-8
  • Molecular Weight : 484.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0422

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0422
Availability details for
Quantity
  • Availability
  • Add to Cart