COM448654066 Combi-Blocks, Inc.

(+/-)-Trans-1,2-bis(2-mercaptoacetamido)cyclohexane

  • MDL number MFCD02684003
  • Cas Number : 257641-01-3
  • Molecular Weight : 262.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0449

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0449
Availability details for
Quantity
  • Availability
  • Add to Cart