COM448654070 Combi-Blocks, Inc.

9,9-Dihexylfluorene

  • MDL number MFCD14584648
  • Cas Number : 123863-97-8
  • Molecular Weight : 334.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0453

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0453
Availability details for
Quantity
  • Availability
  • Add to Cart