COM448654086 Combi-Blocks, Inc.

2-(2-Methoxyethyl)piperidine

  • MDL number MFCD08669774
  • Cas Number : 858523-63-4
  • Molecular Weight : 143.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0469

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0469
Availability details for
Quantity
  • Availability
  • Add to Cart