COM448654095 Combi-Blocks, Inc.

1,3,5-Triphenylisocyanurate

  • MDL number MFCD00367460
  • Cas Number : 1785-02-0
  • Molecular Weight : 357.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0478

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0478
Availability details for
Quantity
  • Availability
  • Add to Cart