COM448654102 Combi-Blocks, Inc.

4-(5-Amino-6-(ethylamino)pyrimidin-4-ylamino)benzamide

  • MDL number MFCD14585497
  • Cas Number : 1206969-06-3
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0485

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0485
Availability details for
Quantity
  • Availability
  • Add to Cart