COM448654103 Combi-Blocks, Inc.

Octafluoroadipoyl chloride

  • MDL number MFCD00054726
  • Cas Number : 336-06-1
  • Molecular Weight : 327.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0486

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0486
Availability details for
Quantity
  • Availability
  • Add to Cart