COM448654106 Combi-Blocks, Inc.

Methyl 3-amino-1-benzylpyrrolidine-3-carboxylate

  • MDL number MFCD09608112
  • Cas Number : 145602-88-6
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0489

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0489
Availability details for
Quantity
  • Availability
  • Add to Cart