COM448654112 Combi-Blocks, Inc.

Pigment red 255

  • MDL number MFCD11870113
  • Cas Number : 54660-00-3
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0495

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0495
Availability details for
Quantity
  • Availability
  • Add to Cart