COM448654117 Combi-Blocks, Inc.

(3-Chloro-5-fluoropyridin-2-yl)methanol

  • MDL number MFCD16607023
  • Cas Number : 1227563-98-5
  • Molecular Weight : 161.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0500

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0500
Availability details for
Quantity
  • Availability
  • Add to Cart