COM448654126 Combi-Blocks, Inc.

4-[(Dimethylamino)methyl]-2,6-dimethylphenol hydrochloride

  • MDL number MFCD00275711
  • Cas Number : 72920-05-9
  • Molecular Weight : 215.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0509

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0509
Availability details for
Quantity
  • Availability
  • Add to Cart