COM448654128 Combi-Blocks, Inc.

1-Iodo-1h,1h-perfluoroheptane

  • MDL number MFCD00233026
  • Cas Number : 212563-43-4
  • Molecular Weight : 460.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0511

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0511
Availability details for
Quantity
  • Availability
  • Add to Cart