COM448654132 Combi-Blocks, Inc.

1-(2-Chlorophenyl)cyclobutanecarbonitrile

  • MDL number MFCD11036689
  • Cas Number : 28049-59-4
  • Molecular Weight : 191.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0516

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0516
Availability details for
Quantity
  • Availability
  • Add to Cart