COM448654133 Combi-Blocks, Inc.

(5-Bromo-2-iodophenyl)methanamine

  • MDL number MFCD06212951
  • Cas Number : 771581-39-6
  • Molecular Weight : 311.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0517

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0517
Availability details for
Quantity
  • Availability
  • Add to Cart