COM448654149 Combi-Blocks, Inc.

2,3-Bis(4-nitrophenyl)-5-phenyltetrazolium chloride hydrate

  • MDL number MFCD09998205
  • Cas Number : 69231-13-6
  • Molecular Weight : 442.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0534

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0534
Availability details for
Quantity
  • Availability
  • Add to Cart