COM448654150 Combi-Blocks, Inc.

Bigchap

  • MDL number MFCD00082541
  • Cas Number : 86303-22-2
  • Molecular Weight : 878.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0535

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0535
Availability details for
Quantity
  • Availability
  • Add to Cart