COM448654154 Combi-Blocks, Inc.

1-Benzhydrylindoline-2,3-dione

  • MDL number MFCD00469414
  • Cas Number : 94878-41-8
  • Molecular Weight : 313.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0539

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0539
Availability details for
Quantity
  • Availability
  • Add to Cart