COM448654174 Combi-Blocks, Inc.

6-Bromo-2-naphthyl-alpha-d-mannopyranoside

  • MDL number MFCD00067621
  • Cas Number : 28541-84-6
  • Molecular Weight : 385.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0560

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0560
Availability details for
Quantity
  • Availability
  • Add to Cart