COM448654180 Combi-Blocks, Inc.

4-(tert-Butoxy)benzotrifluoride

  • MDL number MFCD09763645
  • Cas Number : 16222-44-9
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0566

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0566
Availability details for
Quantity
  • Availability
  • Add to Cart