COM448654193 Combi-Blocks, Inc.

2,4-Dimethyl-5-phenylthiophene

  • MDL number MFCD08276354
  • Cas Number : 57021-49-5
  • Molecular Weight : 188.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0580

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0580
Availability details for
Quantity
  • Availability
  • Add to Cart