COM448654209 Combi-Blocks, Inc.

(4-(4-(4-Methyl-1h-imidazol-1-yl)phenyl)thiazol-2-yl)methanol

  • MDL number MFCD14585382
  • Cas Number : 1206969-30-3
  • Molecular Weight : 271.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0596

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0596
Availability details for
Quantity
  • Availability
  • Add to Cart