COM448654225 Combi-Blocks, Inc.

4-Isobutylcyclohexanecarboxylic acid

  • MDL number MFCD01734735
  • Cas Number : 38792-88-0
  • Molecular Weight : 184.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0612

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0612
Availability details for
Quantity
  • Availability
  • Add to Cart