COM448654231 Combi-Blocks, Inc.

(2-Pyrrolidin-1-ylpyrimidin-5-yl)methanol

  • MDL number MFCD09863240
  • Cas Number : 937796-11-7
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0618

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0618
Availability details for
Quantity
  • Availability
  • Add to Cart