COM448654244 Combi-Blocks, Inc.

2,3,6-Trichlorotoluene

  • MDL number MFCD00013623
  • Cas Number : 2077-46-5
  • Molecular Weight : 195.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0631

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0631
Availability details for
Quantity
  • Availability
  • Add to Cart