COM448654248 Combi-Blocks, Inc.

6-Methoxy-2-methylpyridine-3-carbonitrile

  • MDL number MFCD16659668
  • Cas Number : 105277-11-0
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0635

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0635
Availability details for
Quantity
  • Availability
  • Add to Cart