COM448654264 Combi-Blocks, Inc.

4-(2,3,4,5,6-Pentafluorobenzyloxy)benzyl alcohol

  • MDL number MFCD09749654
  • Cas Number : 471469-89-3
  • Molecular Weight : 304.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0651

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0651
Availability details for
Quantity
  • Availability
  • Add to Cart