COM448654276 Combi-Blocks, Inc.

4-Methylpent-4-enoic acid

  • MDL number MFCD06208588
  • Cas Number : 1001-75-8
  • Molecular Weight : 114.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0664

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0664
Availability details for
Quantity
  • Availability
  • Add to Cart