COM448654278 Combi-Blocks, Inc.

2-Chloro-5-furan-2-yl-pyrazine

  • MDL number MFCD09033268
  • Cas Number : 82619-63-4
  • Molecular Weight : 180.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0666

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0666
Availability details for
Quantity
  • Availability
  • Add to Cart