COM448654297 Combi-Blocks, Inc.

6-Chloro-4-hydroxyquinoline-2-carboxylic acid

  • MDL number MFCD00673835
  • Cas Number : 10174-72-8
  • Molecular Weight : 223.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0685

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0685
Availability details for
Quantity
  • Availability
  • Add to Cart