COM448654303 Combi-Blocks, Inc.

1-[2-(Trifluoromethyl)phenyl]propylamine

  • MDL number MFCD07784288
  • Cas Number : 473732-54-6
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0691

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0691
Availability details for
Quantity
  • Availability
  • Add to Cart