COM448654319 Combi-Blocks, Inc.

2-Bromo-6-methoxyquinoline

  • MDL number MFCD08688605
  • Cas Number : 476161-59-8
  • Molecular Weight : 238.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0707

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0707
Availability details for
Quantity
  • Availability
  • Add to Cart